Carglumic acid 97%
Supplier: Ambeed
|
Warning
|
A304788-100G
A304788-10G
A304788-1G
A304788-250MG
A304788-25G
A304788-500G
A304788-5G
76723-680EA
130
USD
76723-680
76723-682
76723-684
76723-686
76723-688
76723-690
76723-692
Carglumic acid 97%
Carglumic acid
Precaución: for Research Use Only
Formula:
C₆H₁₀N₂O₅ |
MDL Number:
MFCD00047874 CAS Number: 1188-38-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...