Methyl palmitate 98%

Supplier: Ambeed
Warning

Synonyms: Hexadecylic acid methyl ester, Methyl hexadecanoate, Palmitic acid methyl ester, Hexadecanoic acid methyl ester, Palmitic Acid Methyl Ester

A303904-100MG A303904-250MG A303904-25G A303904-5G
76723-662EA 5.31 USD
76723-662 76723-664 76723-666 76723-668
Methyl palmitate 98%
Methyl palmitate

Precaución: for Research Use Only

Formula: C₁₇H₃₄O₂
MW: 270.46 g/mol
MDL Number: MFCD00008994
CAS Number: 112-39-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR