1-(Pyrazin-2-yl)thiourea 97%

Supplier: Ambeed
Danger

A303688-1G A303688-250MG
76723-656EA 13 USD
76723-656 76723-658
1-(Pyrazin-2-yl)thiourea 97%
1-(Pyrazin-2-yl)thiourea

Precaución: for Research Use Only

Formula: C₅H₆N₄S
MDL Number: MFCD04971717
CAS Number: 31437-05-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR