Ethyl-N-methylcarbamate 95%

Supplier: Ambeed
Warning

Synonyms: N-Methylurethane, N-Methylcarbamic Acid Ethyl Ester, Ethyl methylcarbamate

A303654-1G A303654-25G A303654-5G
76723-650EA 13 USD
76723-650 76723-652 76723-654
Ethyl-N-methylcarbamate 95%
Ethyl-N-methylcarbamate

Precaución: for Research Use Only

Formula: C₄H₉NO₂
MDL Number: MFCD00041924
CAS Number: 105-40-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR