4-(Phenylthio)benzene-1,2-diamine 95%

Supplier: Ambeed
Warning

A303537-250MG A303537-5G
76723-646EA 18 USD
76723-646 76723-648
4-(Phenylthio)benzene-1,2-diamine 95%
4-(Phenylthio)benzene-1,2-diamine

Precaución: for Research Use Only

Formula: C₁₂H₁₂N₂S
MDL Number: MFCD01073766
CAS Number: 43156-48-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR