N-(1-Phenylvinyl)acetamide 95%

Supplier: Ambeed
Warning

A301716-100MG A301716-1G A301716-250MG
76723-626EA 41 USD
76723-626 76723-628 76723-630
N-(1-Phenylvinyl)acetamide 95%

Precaución: for Research Use Only

Formula: C₁₀H₁₁NO
MDL Number: MFCD09033632
CAS Number: 57957-24-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR