4-Fluoro-4'-hydroxybiphenyl 98%

Supplier: Ambeed
Warning

Synonyms: 4'-Fluorobiphenyl-1-ol, 4-(4-Fluorophenyl)phenol, 4'-Fluoro-[1,1'-biphenyl]-4-ol

A300367-100G A300367-10G A300367-1G A300367-250MG A300367-25G A300367-5G
76723-606EA 453.01 USD
76723-606 76723-608 76723-610 76723-612 76723-614 76723-616
4-Fluoro-4'-hydroxybiphenyl 98%
4-Fluoro-4'-hydroxybiphenyl

Precaución: for Research Use Only

Formula: C₁₂H₉FO
MW: 188.20 g/mol
MDL Number: MFCD01830385
CAS Number: 324-94-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR