4-Bromo-2-fluorobiphenyl 97%

Supplier: Ambeed
Warning

A299467-100G A299467-10G A299467-25G A299467-5G
76723-598EA 34 USD
76723-598 76723-600 76838-070 77879-588
4-Bromo-2-fluorobiphenyl 97%
4-Bromo-2-fluorobiphenyl

Precaución: for Research Use Only

Formula: C₁₂H₈BrF
MDL Number: MFCD00051716
CAS Number: 41604-19-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR