n-Nonacosane 99%

Supplier: Ambeed
Warning

Synonyms: Nonacosane

A299065-100MG A299065-1G A299065-250MG
76723-592EA 55.17 USD
76723-592 76723-594 76723-596
n-Nonacosane 99%
n-Nonacosane

Precaución: for Research Use Only

Formula: H₃C(CH₂)₂₇CH₃
MW: 408.8 g/mol
MDL Number: MFCD00009356
CAS Number: 630-03-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR