4-(N-((2-Amino-4-oxo-3,4-dihydropteridin-6-yl)methyl)-2,2,2-trifluoroacetamido)benzoic acid 95%

Supplier: Ambeed
Danger

A296648-100MG A296648-1G A296648-250MG
76723-574EA 117 USD
76723-574 76723-576 76723-578
4-(N-((2-Amino-4-oxo-3,4-dihydropteridin-6-yl)methyl)-2,2,2-trifluoroacetamido)benzoic acid 95%
4-(N-((2-Amino-4-oxo-3,4-dihydropteridin-6-yl)methyl)-2,2,2-trifluoroacetamido)benzoic acid

Precaución: for Research Use Only

Formula: C₁₆H₁₁F₃N₆O₄
MDL Number: MFCD00006707
CAS Number: 37793-53-6
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR