1,2,3,4-Tetrahydroacridin-9-amine 98%

Supplier: Ambeed
Danger

A295785-1G A295785-25G A295785-5G
76723-568EA 20 USD
76723-568 76723-570 76723-572
1,2,3,4-Tetrahydroacridin-9-amine 98%
1,2,3,4-Tetrahydroacridin-9-amine

Precaución: for Research Use Only

Formula: C₁₃H₁₄N₂
MDL Number: MFCD00046923
CAS Number: 321-64-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR