1-(3-Nitrophenyl)ethanol 97%

Supplier: Ambeed
Warning

A295417-10G A295417-1G A295417-250MG A295417-25G A295417-5G
76723-546EA 27 USD
76723-546 76723-548 76723-550 76723-552 76723-554
1-(3-Nitrophenyl)ethanol 97%
1-(3-Nitrophenyl)ethanol

Precaución: for Research Use Only

Formula: C₈H₉NO₃
MDL Number: MFCD00053608
CAS Number: 5400-78-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR