N,N′-(1-Methyl-1,2-ethanediyl)bis[3-pyridinecarboxamide] 97%

Supplier: Ambeed
Warning

Synonyms: Nicaraven

A295397-100MG A295397-10MG A295397-1MG A295397-50MG A295397-250MG
76723-538EA 113.52 USD
76723-538 76723-540 76723-542 76723-544 77668-318
N,N′-(1-Methyl-1,2-ethanediyl)bis[3-pyridinecarboxamide] 97%
N,N′-(1-Methyl-1,2-ethanediyl)bis[3-pyridinecarboxamide]

Precaución: for Research Use Only

Formula: C₁₅H₁₆N₄O₂
MW: 284.31 g/mol
MDL Number: MFCD00411904
CAS Number: 79455-30-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR