N-(3-Acetylphenyl)-N-methylacetamide 98%

Supplier: Ambeed
Warning

A290178-1G A290178-250MG A290178-5G
76723-486EA 35 USD
76723-486 76723-488 76723-490
N-(3-Acetylphenyl)-N-methylacetamide 98%
N-(3-Acetylphenyl)-N-methylacetamide

Precaución: for Research Use Only

Formula: C₁₁H₁₃NO₂
MDL Number: MFCD08445602
CAS Number: 325715-13-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR