4-Ethynyl-1,1'-biphenyl 97%

Supplier: Ambeed
Warning

Synonyms: 4-Ethynylbiphenyl, 4-Biphenylacetylene

A289608-100G A289608-10G A289608-1G A289608-250MG A289608-25G A289608-5G
76723-470EA 335 USD
76723-470 76723-472 76723-474 76723-476 76723-478 76723-480
4-Ethynyl-1,1'-biphenyl 97%
4-Ethynyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₀
MDL Number: MFCD00102191
CAS Number: 29079-00-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR