Silver sulphate 99%
Supplier: Ambeed
|
Danger
|
Synonyms:
Disilver sulphate
A289249-10G
A289249-25G
A289249-5G
A289249-500G
76723-464EA
31
USD
76723-464
76723-466
76723-468
77601-286
Silver sulphate 99%
Silver sulphate
Precaución: for Research Use Only
Formula:
Ag₂SO₄ MW: 311.8 g/mol Boiling Pt: 1085 Melting Pt: 655 |
MDL Number:
MFCD00003407 CAS Number: 10294-26-5 UN: 3077 ADR: 9 Ⅲ Merck Index: 13,08603 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...