(S)-2-(Benzylamino)propan-1-ol 95%

Supplier: Ambeed
Warning

A287757-100MG A287757-10G A287757-1G A287757-250MG A287757-25G A287757-5G
76723-420EA 8.49 USD
76723-420 76723-422 76723-424 76723-426 76723-428 76723-430
(S)-2-(Benzylamino)propan-1-ol 95%
(S)-2-(Benzylamino)propan-1-ol

Precaución: for Research Use Only

Formula: C₁₀H₁₅NO
MW: 165.232 g/mol
MDL Number: MFCD03840382
CAS Number: 6940-80-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR