Nicotinic acid N-oxide ≥99%

Supplier: Ambeed
Warning

Synonyms: Oxiniacic Acid, 3-Carboxypyridine 1-oxide

A287612-100G A287612-100MG A287612-25G A287612-5G
76723-412EA 47 USD
76723-412 76723-414 76723-416 77668-274
Nicotinic acid N-oxide ≥99%
Nicotinic acid N-oxide

Precaución: for Research Use Only

Formula: C₆H₅NO₃
MDL Number: MFCD00006201
CAS Number: 2398-81-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR