Methyldiphenylamine 98%

Supplier: Ambeed
Warning

Synonyms: N-Methyl-N-phenylaniline

A286309-1000G A286309-10G A286309-25G A286309-500G A286309-5G
76723-400EA 323.58 USD
76723-400 76723-402 76723-404 76723-406 76723-408
Methyldiphenylamine 98%
Methyldiphenylamine

Precaución: for Research Use Only

Formula: C₁₃H₁₃N
MW: 183.25 g/mol
MDL Number: MFCD00041900
CAS Number: 552-82-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR