1-(2-Methylbenzo[d]oxazol-6-yl)-3-(1,5-naphthyridin-4-yl)urea 99+%
Supplier: Ambeed
|
Warning
|
A284699-10MG
A284699-1MG
A284699-50MG
A284699-5MG
76723-366EA
37.14
USD
76723-366
76723-368
76723-370
76723-372
1-(2-Methylbenzo[d]oxazol-6-yl)-3-(1,5-naphthyridin-4-yl)urea 99+%
1-(2-Methylbenzo[d]oxazol-6-yl)-3-(1,5-naphthyridin-4-yl)urea
Precaución: for Research Use Only
Formula:
C₁₇H₁₃N₅O₂ MW: 319.32 g/mol |
MDL Number:
MFCD06411602 CAS Number: 792173-99-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...