Potassium (4-methylphenyl)trifluoroborate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Potassium trifluoro(p-tolyl)borate, Potassium p-tolyltrifluoroborate, (4-Methylphenyl)trifluoroboric acid potassium salt
A283538-10G
A283538-1G
A283538-250MG
A283538-25G
A283538-5G
76723-348EA
93.36
USD
76723-348
76723-350
76723-352
76723-354
76723-356
Potassium (4-methylphenyl)trifluoroborate 97%
Potassium (4-methylphenyl)trifluoroborate
Precaución: for Research Use Only
Formula:
C₇H₇BF₃K MW: 198.04 g/mol |
MDL Number:
MFCD01318171 CAS Number: 216434-82-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...