2-Quinolinecarbonitrile 97%

Supplier: Ambeed
Warning

Synonyms: Quinoline-2-carbonitrile

A283473-100G A283473-100MG A283473-10G A283473-1G A283473-250MG A283473-25G A283473-5G
76723-334EA 1156.39 USD
76723-334 76723-336 76723-338 76723-340 76723-342 76723-344 76723-346
2-Quinolinecarbonitrile 97%
2-Quinolinecarbonitrile

Precaución: for Research Use Only

Formula: C₁₀H₆N₂
MW: 154.17 g/mol
MDL Number: MFCD00134341
CAS Number: 1436-43-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR