Sebacic acid dihydrazide 96%

Supplier: Ambeed
Danger

Synonyms: Decanedihydrazide

A282570-100G A282570-25G A282570-5G
76723-310EA 66.84 USD
76723-310 76723-312 76723-314
Sebacic acid dihydrazide 96%
Sebacic acid dihydrazide

Precaución: for Research Use Only

Formula: C₁₀H₂₂N₄O₂
MW: 230.3073 g/mol
MDL Number: MFCD00039763
CAS Number: 925-83-7
UN: 1325
ADR: 4.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR