PD 98059 98%

Supplier: Ambeed
Warning

Synonyms: PD98059, 2'-Amino-3'-methoxyflavone, 2-(2-Amino-3-methoxyphenyl)chromone, PD 98059, 2-(2-Amino-3-methoxyphenyl)-4H-1-benzopyran-4-one, 2-(2-Amino-3-methoxyphenyl)-4H-chromen-4-one

A282482-10MG A282482-1MG A282482-50MG A282482-5MG A282482-100MG
76723-302EA 47.75 USD
76723-302 76723-304 76723-306 76723-308 77668-250
PD 98059 98%
PD 98059

Precaución: for Research Use Only

Formula: C₁₆H₁₃NO₃
MW: 267.284 g/mol
MDL Number: MFCD00671789
CAS Number: 167869-21-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR