N-Acetyl-D-mannosamine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Cyclic N-Acetyl-D-mannosamine, N-((2R,3S,4R,5S,6R)-2,4,5-Trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-3-yl)acetamide
A281916-1G
A281916-25G
A281916-5G
A281916-10G
A281916-100G
76723-296EA
15
USD
76723-296
76723-298
76723-300
77601-242
77668-248
N-Acetyl-D-mannosamine 97%
N-Acetyl-D-mannosamine
Precaución: for Research Use Only
Formula:
C₈H₁₅NO₆ MW: 221.21 g/mol (anhy) |
MDL Number:
MFCD00149493 CAS Number: 7772-94-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...