N-Phenyl-m-toluidine 97%

Supplier: Ambeed
Warning

Synonyms: 3-Methyldiphenylamine

A280803-100G A280803-10G A280803-25G
76723-276EA 34 USD
76723-276 76723-278 76723-280
N-Phenyl-m-toluidine 97%
N-Phenyl-m-toluidine

Precaución: for Research Use Only

Formula: C₁₃H₁₃N
MDL Number: MFCD00008530
CAS Number: 1205-64-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR