1,3-Bis(2-hydroxyethyl)urea 97%

Supplier: Ambeed
Warning

A279890-1G A279890-5G A279890-25G
76723-252EA 33.95 USD
76723-252 76723-254 77593-728
1,3-Bis(2-hydroxyethyl)urea 97%
1,3-Bis(2-hydroxyethyl)urea

Precaución: for Research Use Only

Formula: C₅H₁₂N₂O₃
MW: 148.16 g/mol
MDL Number: MFCD00522193
CAS Number: 15438-70-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR