N-(3-Chloropropyl)dibutylamine 95%

Supplier: Ambeed
Warning

Synonyms: N-(3-Chloropropyl)Dibutylamine

A279588-100G A279588-10G A279588-1G A279588-5G
76723-240EA 79 USD
76723-240 76723-242 76723-244 76723-246
N-(3-Chloropropyl)dibutylamine 95%
N-(3-Chloropropyl)dibutylamine

Precaución: for Research Use Only

Formula: C₁₁H₂₄ClN
MDL Number: MFCD13185966
CAS Number: 36421-15-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR