Palladium(II) acetylacetonate 98%

Supplier: Ambeed
Warning

Synonyms: Palladium (II) 2,4-pentanedionate, Bis(2,4-pentanedionato-O,O')palladium (II), Palladium (II) bis(2,4-pentanedionato-O,O'), Palladium(II) 2,4-pentanedionate

A279251-100MG A279251-1G A279251-250MG A279251-5G
76723-226EA 9 USD
76723-226 76723-228 76723-230 76723-232
Palladium(II) acetylacetonate 98%
Palladium(II) acetylacetonate

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₄Pd
MW: 304.64 g/mol
Melting Pt: 190
MDL Number: MFCD00000025
CAS Number: 14024-61-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR