N,N-Bis(2-chloroethyl)-p-toluenesulfonamide 95%

Supplier: Ambeed
Warning

Synonyms: N,N-Bis(2-chloroethyl)-4-methylbenzenesulphonamide

A279052-100G A279052-1G A279052-25G A279052-5G
76723-218EA 154.9 USD
76723-218 76723-220 76723-222 76723-224
N,N-Bis(2-chloroethyl)-p-toluenesulfonamide 95%
N,N-Bis(2-chloroethyl)-p-toluenesulfonamide

Precaución: for Research Use Only

Formula: C₁₁H₁₅Cl₂NO₂S
MW: 296.21 g/mol
MDL Number: MFCD00018944
CAS Number: 42137-88-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR