2-(5-((3-Methylbut-2-en-1-yl)oxy)-2-(3-(4-((3-methylbut-2-en-1-yl)oxy)phenyl)acryloyl)phenoxy)acetic acid 97%

Supplier: Ambeed
Warning

Synonyms: Sofalcone

A278552-10G A278552-1G A278552-1MG A278552-250MG A278552-25G A278552-5G A278552-100G A278552-500G
76723-206EA 69 USD
76723-206 76723-208 76723-210 76723-212 76723-214 76723-216 77668-222 77668-224
2-(5-((3-Methylbut-2-en-1-yl)oxy)-2-(3-(4-((3-methylbut-2-en-1-yl)oxy)phenyl)acryloyl)phenoxy)acetic acid 97%
2-(5-((3-Methylbut-2-en-1-yl)oxy)-2-(3-(4-((3-methylbut-2-en-1-yl)oxy)phenyl)acryloyl)phenoxy)acetic acid

Precaución: for Research Use Only

Formula: C₂₇H₃₀O₆
MDL Number: MFCD00868207
CAS Number: 64506-49-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR