Oxamic hydrazide 98%

Supplier: Ambeed
Danger

Synonyms: Semioxamazide, Amino-oxamide

A278134-1G A278134-25G A278134-5G
76723-196EA 6 USD
76723-196 76723-198 76723-200
Oxamic hydrazide 98%
Oxamic hydrazide

Precaución: for Research Use Only

Formula: C₂H₅N₃O₂
MW: 103.08 g/mol
MDL Number: MFCD00008004
CAS Number: 515-96-8
UN: 1325
ADR: 4.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR