3-Carbamoylphenylboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 2-dioxaborolan-2-yl)-benzamide, 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide, 3-aminocarbonylphenylboronic acid, pinacol ester

A278027-1G A278027-25G A278027-5G
76723-190EA 18 USD
76723-190 76723-192 76723-194
3-Carbamoylphenylboronic acid pinacol ester 98%
3-Carbamoylphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₈BNO₃
MDL Number: MFCD05663886
CAS Number: 188665-74-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR