2-(1-Naphthyl)acetohydrazide 95%
Supplier: Ambeed
|
Warning
|
A277971-10G
A277971-1G
A277971-5G
A277971-25G
76723-186EA
24
USD
76723-186
76723-188
77879-519
77879-518
2-(1-Naphthyl)acetohydrazide 95%
2-(1-Naphthyl)acetohydrazide
Precaución: for Research Use Only
Formula:
C₁₂H₁₂N₂O |
MDL Number:
MFCD00014318 CAS Number: 34800-90-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...