4,4',4''-Trimethoxytrityl alcohol 98+%

Supplier: Ambeed
Warning

Synonyms: Tris(4-methoxyphenyl)methanol

A277839-1G A277839-250MG A277839-25G A277839-5G
76723-178EA 24.41 USD
76723-178 76723-180 76723-182 76723-184
4,4',4''-Trimethoxytrityl alcohol 98+%
4,4',4''-Trimethoxytrityl alcohol

Precaución: for Research Use Only

Formula: C₂₂H₂₂O₄
MW: 350.4077 g/mol
MDL Number: MFCD00014898
CAS Number: 3010-81-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR