Diethyl dipropylmalonate 97%

Supplier: Ambeed
Warning

Synonyms: Diethyl 2,2-dipropylmalonate

A277194-100G A277194-25G A277194-500G A277194-5G
76723-162EA 50 USD
76723-162 76723-164 76723-166 76723-168
Diethyl dipropylmalonate 97%
Diethyl dipropylmalonate

Precaución: for Research Use Only

Formula: C₁₃H₂₄O₄
MDL Number: MFCD00026840
CAS Number: 6065-63-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR