1-Benzenesulfonyl-4-chloro-2-nitrobenzene 95%

Supplier: Ambeed
Warning

Synonyms: 4-Chloro-2-nitro-1-(phenylsulphonyl)benzene, BTB-1

A276415-10MG A276415-1MG A276415-50MG A276415-5MG
76723-144EA 239.77 USD
76723-144 76723-146 76723-148 76723-150
1-Benzenesulfonyl-4-chloro-2-nitrobenzene 95%
1-Benzenesulfonyl-4-chloro-2-nitrobenzene

Precaución: for Research Use Only

Formula: C₁₂H₈ClNO₄S
MW: 297.72 g/mol
MDL Number: MFCD00052250
CAS Number: 86030-08-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR