Sivelestat sodium 98%

Supplier: Ambeed
Warning

Synonyms: ONO-5046, Sodium 2-(2-(4-(pivaloyloxy)phenylsulphonamido)benzamido)acetate tetrahydrate, N-{2-[({4-[2,2-Dimethylpropanoyl)oxy]phenyl}sulphonyl)amino]benzoyl}glycine sodium salt, Sivelestat Sodium Tetrahydrate

A274870-100MG A274870-10MG A274870-1MG A274870-50MG A274870-5MG
76723-124EA 435 USD
76723-124 76723-126 76723-128 76723-130 76723-132
Sivelestat sodium 98%
Sivelestat sodium

Precaución: for Research Use Only

Formula: C₂₀H₂₁N₂NaO₇S·4H₂O
MW: 528.5 g/mol
MDL Number: MFCD01937969
CAS Number: 201677-61-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR