Methyl-4-chlorosalicylate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-chloro-2-hydroxybenzoate

A274340-100G A274340-10G A274340-1G A274340-5G
76723-116EA 344.8 USD
76723-116 76723-118 76723-120 76723-122
Methyl-4-chlorosalicylate 97%
Methyl 4-chlorosalicylate

Precaución: for Research Use Only

Formula: C₈H₇ClO₃
MW: 186.5900 g/mol
MDL Number: MFCD03407436
CAS Number: 22717-55-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR