Octyltrimethoxysilane 98%

Supplier: Ambeed
Warning

Synonyms: Trimethoxy(octyl)silane

A272793-100G A272793-10G A272793-25G A272793-500G
76723-076EA 28.65 USD
76723-076 76723-078 76723-080 76723-082
Octyltrimethoxysilane 98%
Octyltrimethoxysilane

Precaución: for Research Use Only

Formula: C₁₁H₂₆O₃Si
MW: 234.41 g/mol
MDL Number: MFCD00043053
CAS Number: 3069-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR