2-(2-((3,5-Dimethyl-1H-pyrrol-2-yl)methylene)-3-methoxy-2H-pyrrol-5-yl)-1H-indole ≥99%

Supplier: Ambeed
Warning

Synonyms: Obatoclax

A272644-1MG A272644-2MG
76723-064EA 18.04 USD
76723-064 76723-066
2-(2-((3,5-Dimethyl-1H-pyrrol-2-yl)methylene)-3-methoxy-2H-pyrrol-5-yl)-1H-indole ≥99%
2-(2-((3,5-Dimethyl-1H-pyrrol-2-yl)methylene)-3-methoxy-2H-pyrrol-5-yl)-1H-indole

Precaución: for Research Use Only

Formula: C₂₀H₁₉N₃O
MW: 317.38 g/mol
MDL Number: MFCD09833233
CAS Number: 803712-67-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR