Ethyl-4-hydroxy-8-(trifluoromethyl)quinoline-3-carboxylate 96%

Supplier: Ambeed
Warning

Synonyms: Ethyl 4-hydroxy-8-(trifluoromethyl)quinoline-3-carboxylate

A271917-10G A271917-1G A271917-5G
76723-046EA 33.95 USD
76723-046 76723-048 76723-050
Ethyl-4-hydroxy-8-(trifluoromethyl)quinoline-3-carboxylate 96%
Ethyl 4-hydroxy-8-(trifluoromethyl)quinoline-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₁₀F₃NO₃
MW: 285.2186 g/mol
MDL Number: MFCD09878378
CAS Number: 23851-84-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR