Perillartine 98%

Supplier: Ambeed
Warning

Synonyms: (NZ)-N-[(4-prop-1-en-2-ylcyclohexen-1-yl)methylidene]hydroxylamine, (E)-4-(Prop-1-en-2-yl)cyclohex-1-enecarbaldehyde oxime

A269944-100G A269944-100MG A269944-10G A269944-25G
76723-032EA 98.67 USD
76723-032 76723-034 76723-036 76723-038
Perillartine 98%
Perillartine

Precaución: for Research Use Only

Formula: C₁₀H₁₅NO
MW: 165.232 g/mol
MDL Number: MFCD00019421
CAS Number: 30950-27-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR