N-(Cyclohexylthio)phthalimide 95%

Supplier: Ambeed
Warning

Synonyms: 2-(Cyclohexylthio)isoindoline-1,3-dione, Cyclohexyl N-Phthalimidyl Sulphide

A269022-1000G A269022-100G A269022-10G A269022-500G A269022-25G
76723-004EA 149 USD
76723-004 76723-006 76723-008 76723-010 77092-608
N-(Cyclohexylthio)phthalimide 95%
N-(Cyclohexylthio)phthalimide

Precaución: for Research Use Only

Formula: C₁₄H₁₅NO₂S
MDL Number: MFCD00072247
CAS Number: 17796-82-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR