(S)-6-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-(((benzyloxy)carbonyl)amino)hexanoic acid 95%

Supplier: Ambeed
Warning

Synonyms: Z-Lys(Fmoc)-OH, Cbz-Lys(Fmoc)-OH

A268890-1G A268890-25G A268890-5G
76723-002EA 10.61 USD
76723-002 77668-168 77668-170
(S)-6-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-(((benzyloxy)carbonyl)amino)hexanoic acid 95%
(S)-6-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-(((benzyloxy)carbonyl)amino)hexanoic acid

Precaución: for Research Use Only

Formula: C₂₉H₃₀N₂O₆
MW: 502.567 g/mol
MDL Number: MFCD00065662
CAS Number: 105751-18-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR