2-Phenyl-5-benzimidazolesulfonic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ensulizole, Phenylbenzimidazole sulphonic acid, 1H-Benzimidazole-5-sulphonic acid
A268828-1000G
A268828-100G
A268828-25G
A268828-500G
76722-994EA
123.07
USD
76722-994
76722-996
76722-998
76723-000
2-Phenyl-5-benzimidazolesulfonic acid 98%
2-Phenyl-5-benzimidazolesulphonic acid
Precaución: for Research Use Only
Formula:
C₁₃H₁₀N₂O₃S MW: 274.29 g/mol |
MDL Number:
MFCD00053007 CAS Number: 27503-81-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...