Ethyl-4-piperidinecarboxylate hydrochloride 97%

Supplier: Ambeed
Warning

A268815-100G A268815-25G
76722-992EA 20.16 USD
76722-992 77668-166
Ethyl-4-piperidinecarboxylate hydrochloride 97%
Ethyl 4-piperidinecarboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₆ClNO₂
MDL Number: MFCD06655993
CAS Number: 147636-76-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR