(2-Amino-5-nitrophenyl)(2-fluorophenyl)methanone 97%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-2-Fluoro-5-Nitrobenzophenone

A267809-100G A267809-1G A267809-5G
76722-970EA 245.07 USD
76722-970 76722-972 76722-974
(2-Amino-5-nitrophenyl)(2-fluorophenyl)methanone 97%
(2-Amino-5-nitrophenyl)(2-fluorophenyl)methanone

Precaución: for Research Use Only

Formula: C₁₃H₉FN₂O₃
MW: 260.22 g/mol
MDL Number: MFCD00970341
CAS Number: 344-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR