N-(n-propyl)acetamide 98%

Supplier: Ambeed
Warning

A265790-1G A265790-250MG A265790-5G
76722-932EA 80 USD
76722-932 76722-934 76722-936
N-(n-propyl)acetamide 98%
N-(n-propyl)acetamide

Precaución: for Research Use Only

Formula: C₅H₁₁NO
MDL Number: MFCD00051554
CAS Number: 5331-48-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR