N-Bromosaccharin 98%

Supplier: Ambeed
Warning

A265432-1G A265432-5G
76722-924EA 54 USD
76722-924 76722-926
N-Bromosaccharin 98%
N-Bromosaccharin

Precaución: for Research Use Only

Formula: C₇H₄BrNO₃S
MDL Number: MFCD03844772
CAS Number: 35812-01-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR